CH4415172896 ChemBridge Corporation

1-(2-methyl-4-pyridinyl)piperazine

  • MDL number MFCD08691575
  • Cas Number : 98010-38-9
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4087866

Safety Information

MSDS https://www.hit2lead.com/sds/4087866_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart