CH4415172906 ChemBridge Corporation

(1-isopropyl-1H-imidazol-5-yl)methanol

  • MDL number MFCD08691606
  • Cas Number : 80304-46-7
  • Molecular Weight : 140.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087876

Safety Information

MSDS https://www.hit2lead.com/sds/4087876_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart