CH4415172907 ChemBridge Corporation

2-(chloromethyl)-4-methoxypyridine

  • MDL number MFCD07774101
  • Cas Number : 99651-28-2
  • Molecular Weight : 157.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087877

Safety Information

MSDS https://www.hit2lead.com/sds/4087877_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart