CH4415172909 ChemBridge Corporation

tert-butyl 3-(aminomethyl)-3-methyl-1-azetidinecarboxylate

  • MDL number MFCD12408560
  • Cas Number : 1158758-85-0
  • Molecular Weight : 200.278

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4046397

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4046397
Availability details for
Quantity
  • Availability
  • Add to Cart