CH4415172919 ChemBridge Corporation

3-acetyl-2(1H)-pyridinone

  • MDL number MFCD11977425
  • Cas Number : 62838-65-7
  • Molecular Weight : 137.136

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4048059

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4048059
Availability details for
Quantity
  • Availability
  • Add to Cart