CH4415172921 ChemBridge Corporation

2-(4-chlorophenyl)-2-(1-piperidinyl)ethanamine

  • MDL number MFCD08667748
  • Cas Number : 914207-57-1
  • Molecular Weight : 238.7564

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://www.hit2lead.com/building-blocks/4087892

Safety Information

msds https://www.hit2lead.com/building-blocks/4087892
Availability details for
Quantity
  • Availability
  • Add to Cart