CH4415172922 ChemBridge Corporation

6-amino-1,3-dihydro-2H-indol-2-one hydrochloride

  • MDL number MFCD15147389
  • Cas Number : 101389-22-4
  • Molecular Weight : 184.6229

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://www.hit2lead.com/building-blocks/4087893

Safety Information

msds https://www.hit2lead.com/building-blocks/4087893
Availability details for
Quantity
  • Availability
  • Add to Cart