CH4415172926 ChemBridge Corporation

2,6-dichloro-4-pyrimidinecarbonitrile

  • MDL number MFCD16988336
  • Cas Number : 26293-93-6
  • Molecular Weight : 173.9875

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4048582

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4048582
Availability details for
Quantity
  • Availability
  • Add to Cart