CH4415172930 ChemBridge Corporation

[2-(4-morpholinylcarbonyl)phenyl]amine hydrochloride

  • MDL number MFCD00136428
  • Cas Number : 1048648-81-2
  • Molecular Weight : 242.702

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087901

Safety Information

MSDS https://www.hit2lead.com/sds/4087901_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart