CH4415172944 ChemBridge Corporation

1-(5-cyclopropyl-3-isoxazolyl)methanamine

  • MDL number MFCD11053974
  • Cas Number : 1060817-49-3
  • Molecular Weight : 138.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087915

Safety Information

MSDS https://www.hit2lead.com/sds/4087915_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart