CH4415172945 ChemBridge Corporation

(2-amino-4-chlorophenyl)dimethylamine

  • MDL number MFCD06655107
  • Cas Number : 183251-88-9
  • Molecular Weight : 170.6393

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087916

Safety Information

MSDS https://www.hit2lead.com/sds/4087916_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart