CH4415172946 ChemBridge Corporation

4-[(dimethylamino)methyl]-4-piperidinol

  • MDL number MFCD10686624
  • Cas Number : 695145-47-2
  • Molecular Weight : 158.2413

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087917

Safety Information

MSDS https://www.hit2lead.com/sds/4087917_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart