CH4415172947 ChemBridge Corporation

(2-amino-4-chloro-6-methylphenyl)amine

  • MDL number MFCD03426174
  • Cas Number : 109671-52-5
  • Molecular Weight : 156.6128

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087918

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4087918
Availability details for
Quantity
  • Availability
  • Add to Cart