CH4415172950 ChemBridge Corporation

6-(4-methyl-1-piperazinyl)-3-pyridinamine hydrochloride

  • MDL number MFCD11052569
  • Cas Number : 1234971-67-5
  • Molecular Weight : 228.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data HCl
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087921

Safety Information

MSDS https://www.hit2lead.com/sds/4087921_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart