CH4415172957 ChemBridge Corporation

N-benzylguanidine acetate

  • MDL number MFCD19981491
  • Cas Number : 117053-38-0
  • Molecular Weight : 209.245

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CH3COOH
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087928

Safety Information

MSDS https://www.hit2lead.com/sds/4087928_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart