CH4415172968 ChemBridge Corporation

2-bromo-4,5-dimethyl-1,3-thiazole hydrobromide

  • MDL number MFCD18071181
  • Cas Number : 1559060-02-4
  • Molecular Weight : 272.9888

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087939

Safety Information

MSDS https://www.hit2lead.com/sds/4087939_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart