CH4415172980 ChemBridge Corporation

N-methyl-1-(2-thienyl)methanamine hydrochloride

  • MDL number MFCD03306020
  • Cas Number : 7404-67-3
  • Molecular Weight : 163.6683

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087952

Safety Information

MSDS https://www.hit2lead.com/sds/4087952_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart