CH4415172981 ChemBridge Corporation

2-(2-furyl)ethanamine

  • MDL number MFCD04113973
  • Cas Number : 1121-46-6
  • Molecular Weight : 111.1418

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087953

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4087953
Availability details for
Quantity
  • Availability
  • Add to Cart