CH4415172982 ChemBridge Corporation

(1,1-dioxidotetrahydro-3-thienyl)amine

  • MDL number MFCD00022545
  • Cas Number : 6338-70-1
  • Molecular Weight : 135.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087954

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/4087954_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart