CH4415172991 ChemBridge Corporation

2-(2-pyrrolidinyl)-1H-benzimidazole

  • MDL number MFCD01859287
  • Cas Number : 638141-64-7
  • Molecular Weight : 187.241

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087963

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4087963
Availability details for
Quantity
  • Availability
  • Add to Cart