CH4415172992 ChemBridge Corporation

(1H-imidazol-2-ylmethyl)amine hydrochloride

  • MDL number MFCD07366514
  • Cas Number : 138799-95-8
  • Molecular Weight : 133.5794

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087964

Safety Information

MSDS https://www.hit2lead.com/sds/4087964_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart