CH4415172994 ChemBridge Corporation

N-methyl-2-(1H-pyrazol-4-yl)ethanamine

  • MDL number MFCD06751858
  • Cas Number : 956949-79-4
  • Molecular Weight : 125.1716

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087966

Safety Information

MSDS https://www.hit2lead.com/sds/4087966_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart