CH4415173003 ChemBridge Corporation

2,3,4,5-tetrahydro-1,4-benzoxazepine

  • MDL number MFCD08669765
  • Cas Number : 17775-01-8
  • Molecular Weight : 149.1897

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087976

Safety Information

MSDS https://www.hit2lead.com/sds/4087976_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart