CH4415173016 ChemBridge Corporation

N,N-dimethyl-2-(4-piperidinyl)ethanamine

  • MDL number MFCD00047332
  • Cas Number : 102308-48-5
  • Molecular Weight : 156.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087989

Safety Information

MSDS https://www.hit2lead.com/sds/4087989_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart