CH4415173026 ChemBridge Corporation

1,3-benzothiazole-2-carboxylic acid

  • MDL number MFCD02682025
  • Molecular Weight : 179.1958

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087999

Safety Information

MSDS https://www.hit2lead.com/sds/4087999_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart