CH4415173029 ChemBridge Corporation

2-(1H-pyrazol-1-yl)ethanamine

  • MDL number MFCD03727230
  • Cas Number : 101395-71-5
  • Molecular Weight : 111.1451

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087747

Safety Information

MSDS https://www.hit2lead.com/sds/4087747_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart