CH4415173037 ChemBridge Corporation

1-methyl-1H-benzimidazol-6-amine

  • MDL number MFCD09764050
  • Cas Number : 26530-93-8
  • Molecular Weight : 147.1772

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4088011

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4088011
Availability details for
Quantity
  • Availability
  • Add to Cart