CH4424844936 ChemBridge Corporation

[2-(6-chloro-5-methyl-1,3-benzoxazol-2-yl)ethyl]amine hydrochloride

  • MDL number MFCD11696375
  • Cas Number : 1158324-46-9
  • Molecular Weight : 247.1211

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3002156

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3002156
Availability details for
Quantity
  • Availability
  • Add to Cart