CH4424844940 ChemBridge Corporation

{2-[1-(methylsulfonyl)-3-piperidinyl]ethyl}amine hydrochloride

  • MDL number MFCD12027120
  • Cas Number : 1185302-17-3
  • Molecular Weight : 242.7667

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3002303

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3002303
Availability details for
Quantity
  • Availability
  • Add to Cart