CH4424844951 ChemBridge Corporation

[4-(2-pyridinylmethyl)phenyl]amine dihydrochloride

  • MDL number MFCD08070700
  • Cas Number : 96616-23-8
  • Molecular Weight : 257.159

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3002671

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3002671
Availability details for
Quantity
  • Availability
  • Add to Cart