CH4424844971 ChemBridge Corporation

[2-(4-biphenylyloxy)ethyl]amine hydrochloride

  • MDL number MFCD08445694
  • Cas Number : 1051368-98-9
  • Molecular Weight : 249.7359

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3005254

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3005254
Availability details for
Quantity
  • Availability
  • Add to Cart