CH4424844975 ChemBridge Corporation

{3-[(2-isopropyl-1H-imidazol-1-yl)methyl]phenyl}amine dihydrochloride

  • MDL number MFCD18205897
  • Cas Number : 1211449-75-0
  • Molecular Weight : 288.2161

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3005409

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3005409
Availability details for
Quantity
  • Availability
  • Add to Cart