CH4424845095 ChemBridge Corporation

2-(chloromethyl)-3-methylpyridine hydrochloride

  • MDL number MFCD07781109
  • Cas Number : 4370-22-3
  • Molecular Weight : 178.0591

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4002155

Safety Information

MSDS https://www.hit2lead.com/sds/4002155_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart