CH4424845121 ChemBridge Corporation

{1-[1-(2-fluorophenyl)-1H-pyrazol-4-yl]ethyl}amine hydrochloride

  • MDL number MFCD09431461
  • Cas Number : 1049791-50-5
  • Molecular Weight : 241.6924

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4002416

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4002416
Availability details for
Quantity
  • Availability
  • Add to Cart