CH4424845129 ChemBridge Corporation

[(3-benzyl-1,2,4-oxadiazol-5-yl)methyl]amine hydrochloride

  • MDL number MFCD09997606
  • Cas Number : 1185301-35-2
  • Molecular Weight : 225.6748

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4002480

Safety Information

MSDS https://www.hit2lead.com/sds/4002480_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart