CH4424845161 ChemBridge Corporation

6-isobutyl-2-methyl-4-pyrimidinecarboxylic acid hydrochloride

  • MDL number MFCD09864224
  • Cas Number : 1609400-12-5
  • Molecular Weight : 230.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data HCl
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4002751

Safety Information

MSDS https://www.hit2lead.com/sds/4002751_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart