CH4424845179 ChemBridge Corporation

4-(benzyloxy)piperidine hydrochloride

  • MDL number MFCD07368306
  • Cas Number : 81151-68-0
  • Molecular Weight : 227.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4002977

Safety Information

MSDS https://www.hit2lead.com/sds/4002977_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart