CH4424845201 ChemBridge Corporation

4-amino-1-adamantanol hydrochloride

  • MDL number MFCD01825646
  • Cas Number : 20098-19-5
  • Molecular Weight : 203.709

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003083

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4003083
Availability details for
Quantity
  • Availability
  • Add to Cart