CH4424845209 ChemBridge Corporation

(2-isopropoxyphenyl)amine hydrochloride

  • MDL number MFCD10687232
  • Cas Number : 748771-95-1
  • Molecular Weight : 187.6666

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003147

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4003147
Availability details for
Quantity
  • Availability
  • Add to Cart