CH4424845217 ChemBridge Corporation

1-benzyl-2-piperidinecarboxylic acid hydrochloride hydrate

  • MDL number MFCD18483368
  • Molecular Weight : 273.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data HCl H2O
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003219

Safety Information

MSDS https://www.hit2lead.com/sds/4003219_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart