CH4424845218 ChemBridge Corporation

(8-methyl-5-quinolinyl)boronic acid hydrate

  • MDL number MFCD13193789
  • Molecular Weight : 205.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data H2O
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003227

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/4003227_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart