CH4424845224 ChemBridge Corporation

[4-(1H-pyrazol-1-yl)benzyl]amine hydrochloride

  • MDL number MFCD11505500
  • Cas Number : 1107632-13-2
  • Molecular Weight : 209.6754

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003287

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4003287
Availability details for
Quantity
  • Availability
  • Add to Cart