CH4424845241 ChemBridge Corporation

2-(3-piperidinyl)-1,3-benzothiazole hydrochloride

  • MDL number MFCD08447141
  • Cas Number : 51785-16-1
  • Molecular Weight : 254.7789

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003384

Safety Information

MSDS https://www.hit2lead.com/sds/4003384_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart