CH4424845246 ChemBridge Corporation

[(4-methoxyphenyl)(4-pyridinyl)methyl]amine dihydrochloride

  • MDL number MFCD06800958
  • Cas Number : 1185300-09-7
  • Molecular Weight : 287.1849

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003415

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4003415
Availability details for
Quantity
  • Availability
  • Add to Cart