CH4424845261 ChemBridge Corporation

2,5,7-trimethyl-4-quinolinecarboxylic acid hydrate

  • MDL number MFCD13193831
  • Cas Number : 1609403-65-7
  • Molecular Weight : 233.2631

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data H2O
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003472

Safety Information

MSDS https://www.hit2lead.com/sds/4003472_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart