CH4424845271 ChemBridge Corporation

(1H-tetrazol-5-ylmethyl)amine hydrochloride

  • MDL number MFCD11857773
  • Cas Number : 118764-13-9
  • Molecular Weight : 135.5555

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003550

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4003550
Availability details for
Quantity
  • Availability
  • Add to Cart