CH4424845277 ChemBridge Corporation

rac-(1R,2S)-2-propylcyclopropanecarboxylic acid

  • MDL number MFCD16039693
  • Cas Number : 1392481-20-7
  • Molecular Weight : 128.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003600

Safety Information

MSDS https://www.hit2lead.com/sds/4003600_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart