CH4424845288 ChemBridge Corporation

[2-(6-fluoro-1H-benzimidazol-2-yl)ethyl]amine hydrochloride

  • MDL number MFCD11841214
  • Cas Number : 1177315-68-2
  • Molecular Weight : 215.6551

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003825

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4003825
Availability details for
Quantity
  • Availability
  • Add to Cart