CH4424845327 ChemBridge Corporation

1-(2-methoxybenzoyl)piperazine trifluoroacetate

  • MDL number MFCD06800954
  • Cas Number : 436099-85-3
  • Molecular Weight : 334.291

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4004080

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4004080
Availability details for
Quantity
  • Availability
  • Add to Cart