CH4424845332 ChemBridge Corporation

[(3-methyl-2-thienyl)methyl]amine hydrochloride

  • MDL number MFCD09971550
  • Cas Number : 1071674-80-0
  • Molecular Weight : 163.6683

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4004093

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4004093
Availability details for
Quantity
  • Availability
  • Add to Cart